Assay Details
Target Gene Details
Entrez Gene ID: | 69270 |
Gene Name: | GINS complex subunit 1 (Psf1 homolog) |
Gene Aliases: |
2810418N01Rik, Gins4, PSF1, mKIAA0186 |
Location: |
Chr.2:150909557-150931280 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gins1 | NM_001163476.1 | NP_001156948.1 | ||
| NM_027014.1 | NP_081290.1 | |||
| AK013116.1 | BAB28658.1 | |||
| AK129081.1 | BAC97891.1 | |||
| BC027537.1 | AAH27537.1 | |||
| BQ553304.1 |
More Information
Set Membership: |
|

Genomic Map