Assay Details
Target Gene Details
Entrez Gene ID: | 14425 |
Gene Name: | UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 3 |
Gene Aliases: |
- |
Location: |
Chr.2:66082766-66124793 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Galnt3 | NM_015736.2 | NP_056551.2 | ||
| AK019995.1 | ||||
| AK135489.1 | BAE22551.1 | |||
| BC043331.1 | AAH43331.1 | |||
| U70538.1 | AAB09579.1 |
More Information
Set Membership: |
|

Genomic Map