Assay Details
Target Gene Details
Entrez Gene ID: | 241391 |
Gene Name: | UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 5 |
Gene Aliases: |
4832424J23 |
Location: |
Chr.2:57997264-58041027 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Galnt5 | NM_172855.3 | NP_766443.2 | ||
| BC116332.1 |
More Information
Set Membership: |
|

Genomic Map