Assay Details
Target Gene Details
Entrez Gene ID: | 383766 |
Gene Name: | TBC/LysM associated domain containing 2 |
Gene Aliases: |
Gm1332 |
Location: |
Chr.2:157075251-157096948 on Build GRCm38 |
Assay Gene Location: | Overlaps - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tldc2 | NM_001177439.1 | NP_001170910.1 |

Genomic Map