Assay Details
Target Gene Details
Entrez Gene ID: | 70231 |
Gene Name: | golgi reassembly stacking protein 2 |
Gene Aliases: |
0610011A07Rik, 2410043M02Rik, 5730520M13Rik, 9430094F20Rik, AW552058, GOLPH2, GRASP55, GRS2, p59 |
Location: |
Chr.2:70661489-70691725 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gorasp2 | NM_027352.4 | NP_081628.3 | ||
| NR_027343.1 | ||||
| AK002515.1 | ||||
| AK010664.2 | BAE43220.1 | |||
| AK033413.1 | BAC28276.1 | |||
| AK077153.1 | ||||
| AK077683.1 | BAC36954.1 | |||
| AK077706.1 | BAC36969.1 | |||
| AK088239.1 | ||||
| AK148337.1 | BAE28492.1 | |||
| AK151843.1 | BAE30735.1 | |||
| AK152475.1 | BAE31249.1 | |||
| AK152544.1 | BAE31299.1 | |||
| AK152603.1 | BAE31351.1 | |||
| AK153049.1 | BAE31677.1 | |||
| AK153555.1 | ||||
| AK159469.1 | BAE35110.1 | |||
| AK167823.1 | BAE39845.1 | |||
| AK168928.1 | BAE40741.1 | |||
| AK169382.1 | BAE41129.1 | |||
| AK179912.1 | ||||
| AK184024.1 | ||||
| AK188100.1 | ||||
| AK197786.1 | ||||
| AK198884.1 | ||||
| AK207289.1 | ||||
| AK216502.1 | ||||
| AK218436.1 | ||||
| AK219116.1 | ||||
| BC005600.1 | AAH05600.1 | |||
| BC007468.1 | AAH07468.1 | |||
| BC016455.1 | AAH16455.1 |

Genomic Map