Assay Details
Target Gene Details
Entrez Gene ID: | 545481 |
Gene Name: | Rho GTPase activating protein 40 |
Gene Aliases: |
C20orf95, Gm14203, OTTMUSG00000015933 |
Location: |
Chr.2:158512796-158550628 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arhgap40 | NM_001145015.1 | NP_001138487.1 |
More Information
Set Membership: |
|

Genomic Map