Assay Details
Target Gene Details
Entrez Gene ID: | 229004 |
Gene Name: | glucocorticoid modulatory element binding protein 2 |
Gene Aliases: |
AI839884 |
Location: |
Chr.2:181251449-181287974 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gmeb2 | NM_198169.2 | 6 | 666 | NP_937812.1 |
| AB221583.1 | 5 | 546 | ||
| AB221584.1 | 5 | 552 | ||
| AK129319.1 | 2 | 198 | BAC98129.2 | |
| BC029763.1 | 5 | 489 | AAH29763.1 | |
| BC059066.1 | 6 | 666 | AAH59066.1 |

Genomic Map