Assay Details
Target Gene Details
Entrez Gene ID: | 99412 |
Gene Name: | golgi autoantigen, golgin subfamily a, 2 |
Gene Aliases: |
AW555139, GM130, mKIAA4150 |
Location: |
Chr.2:32288221-32307921 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Golga2 | NM_001080968.1 | NP_001074437.1 | ||
| NM_133852.2 | NP_598613.2 | |||
| AK076370.1 | 6 | 514 | ||
| AK077516.1 | ||||
| AK220509.1 | BAD90300.1 | |||
| BC011407.1 | AAH11407.1 |
More Information
Set Membership: |
|

Genomic Map