Assay Details
Target Gene Details
Entrez Gene ID: | 170718 |
Gene Name: | isocitrate dehydrogenase 3 (NAD+) beta |
Gene Aliases: |
C78231 |
Location: |
Chr.2:130279309-130284542 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Idh3b | NM_130884.4 | NP_570954.1 | ||
| AW049896.1 | ||||
| AY035212.1 | AAK64606.1 | |||
| BC009022.1 | AAH09022.1 | |||
| CN456483.1 |
More Information
Set Membership: |
|

Genomic Map