Assay Details
Target Gene Details
Entrez Gene ID: | 14810 |
Gene Name: | glutamate receptor, ionotropic, NMDA1 (zeta 1) |
Gene Aliases: |
GluN1, GluRdelta1, GluRzeta1, M100174, NMD-R1, NMDAR1, NR1, Nmdar, Rgsc174 |
Location: |
Chr.2:25291177-25319198 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Grin1 | NM_001177656.2 | NP_001171127.1 | ||
| NM_001177657.2 | NP_001171128.1 | |||
| NM_008169.3 | NP_032195.1 | |||
| AK158140.1 | BAE34379.1 | |||
| AK158175.1 | ||||
| BC039157.1 | AAH39157.1 | |||
| D10028.1 | BAA00920.1 |
More Information
Set Membership: |
|

Genomic Map