Assay Details
Target Gene Details
Entrez Gene ID: | 52856 |
Gene Name: | mitochondrial ribosome associated GTPase 2 |
Gene Aliases: |
1810011P19Rik, 2900056P18Rik, D2Bwg0647e, Gtpbp5 |
Location: |
Chr.2:180070593-180085902 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mtg2 | NM_001083328.1 | 7 | 831 | NP_001076797.1 |
| NM_001083329.1 | 6 | 737 | NP_001076798.1 | |
| NM_028422.1 | 7 | 876 | NP_082698.1 | |
| NM_181424.3 | 8 | 970 | NP_852089.2 | |
| AK007444.1 | 5 | 823 | ||
| AK040689.1 | 7 | 1360 | ||
| AK183355.1 | ||||
| BC028518.1 | AAH28518.1 | |||
| DT929445.1 | 4 | 454 |

Genomic Map