Assay Details
Target Gene Details
Entrez Gene ID: | 229279 |
Gene Name: | heterogeneous nuclear ribonucleoprotein A3 |
Gene Aliases: |
2410013L13Rik, 2610209F03Rik, 2610510D13Rik, Hnrpa3 |
Location: |
Chr.2:75659230-75669407 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hnrnpa3 | NM_053263.1 | NP_444493.1 | ||
| NM_146130.3 | NP_666242.2 | |||
| NM_198090.2 | NP_932758.1 | |||
| AB201711.1 | BAD89508.1 | |||
| AF463524.1 | AAN76992.1 | |||
| AK133860.1 | BAE21894.1 | |||
| AK169663.1 | BAE41285.1 | |||
| BC023828.1 | AAH23828.1 | |||
| BC023908.1 | AAH23908.1 | |||
| BC038364.1 | AAH38364.1 | |||
| BC057655.1 | AAH57655.1 | |||
| BC062198.1 | AAH62198.1 | |||
| BC064824.1 | AAH64824.1 | |||
| BC085474.1 | AAH85474.1 | |||
| BC092083.1 | AAH92083.1 | |||
| BC116250.1 | ||||
| BC116251.1 | ||||
| BC157919.1 | ||||
| BC158038.1 | ||||
| CO045618.1 |
More Information
Set Membership: |
|

Genomic Map