Assay Details
Target Gene Details
Entrez Gene ID: | 64436 |
Gene Name: | inositol polyphosphate-5-phosphatase E |
Gene Aliases: |
1200002L24Rik, 72kDa, mKIAA0123 |
Location: |
Chr.2:26396249-26409219 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Inpp5e | NM_001290437.1 | 7 | 1694 | NP_001277366.1 |
| NM_033134.3 | 6 | 1978 | NP_149125.1 | |
| NR_110957.1 | 6 | 2065 | ||
| AF226683.1 | 6 | 1940 | AAF86957.1 | |
| AK047245.1 | 5 | 1843 | ||
| AK080075.1 | 6 | 1960 | BAC37823.1 | |
| AK085439.1 | 7 | 1698 | ||
| AK154097.1 | 6 | 1979 | BAE32374.1 | |
| AK158135.1 | 6 | 1963 | BAE34376.1 | |
| AK170174.1 | 7 | 1984 | ||
| AK170786.1 | 6 | 1935 | BAE42028.1 | |
| AK170812.1 | 6 | 1949 | ||
| BC052717.1 | 6 | 1947 | AAH52717.1 | |
| BC080295.1 | 6 | 1943 | AAH80295.1 |
More Information
Set Membership: |
|

Genomic Map