Assay Details
Target Gene Details
Entrez Gene ID: | 64436 |
Gene Name: | inositol polyphosphate-5-phosphatase E |
Gene Aliases: |
1200002L24Rik, 72kDa, mKIAA0123 |
Location: |
Chr.2:26396249-26409219 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Inpp5e | NM_001290437.1 | NP_001277366.1 | ||
| NM_033134.3 | NP_149125.1 | |||
| NR_110957.1 | ||||
| AF226683.1 | AAF86957.1 | |||
| AK047245.1 | ||||
| AK080075.1 | BAC37823.1 | |||
| AK085439.1 | ||||
| AK154097.1 | BAE32374.1 | |||
| AK158135.1 | BAE34376.1 | |||
| AK170174.1 | ||||
| AK170786.1 | BAE42028.1 | |||
| AK170812.1 | ||||
| BC052717.1 | AAH52717.1 | |||
| BC080295.1 | AAH80295.1 |
More Information
Set Membership: |
|

Genomic Map