Assay Details
Target Gene Details
Entrez Gene ID: | 59091 |
Gene Name: | junctophilin 2 |
Gene Aliases: |
1110002E14Rik, JP-2, Jp2 |
Location: |
Chr.2:163336242-163398049 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Jph2 | NM_001205076.1 | NP_001192005.1 | ||
| NM_021566.2 | NP_067541.1 | |||
| AB024447.1 | BAB12044.1 | |||
| AK157145.1 | BAE33978.1 |

Genomic Map