Assay Details
Target Gene Details
Entrez Gene ID: | 16176 |
Gene Name: | interleukin 1 beta |
Gene Aliases: |
IL-1beta, Il-1b |
Location: |
Chr.2:129363843-129371173 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il1b | NM_008361.3 | NP_032387.1 | ||
| AK156396.1 | ||||
| AK157245.1 | ||||
| AK168047.1 | BAE40029.1 | |||
| AK224980.1 | ||||
| AK225002.1 | ||||
| BC011437.1 | AAH11437.1 | |||
| M15131.1 | AAA39276.1 |
More Information
Set Membership: |
|

Genomic Map