Assay Details
Target Gene Details
Entrez Gene ID: | 215274 |
Gene Name: | interleukin 1 family, member 10 |
Gene Aliases: |
- |
Location: |
Chr.2:24287095-24293820 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il1f10 | NM_153077.2 | NP_694717.1 | ||
| AY071844.1 | AAL67155.1 | |||
| BC107311.1 | AAI07312.1 | |||
| BC107312.1 | AAI07313.1 |
More Information
Set Membership: |
|

Genomic Map