Assay Details
Target Gene Details
Entrez Gene ID: | 54448 |
Gene Name: | interleukin 1 family, member 6 |
Gene Aliases: |
Fil1, IL-1H1, IL1RP2, Il1f9, Il36a |
Location: |
Chr.2:24215417-24225701 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il1f6 | NM_019450.3 | NP_062323.1 | ||
| AF200493.1 | AAF69249.1 | |||
| AF206697.1 | AAG35671.1 | |||
| AK004061.1 | BAB23147.1 | |||
| AK224928.1 | ||||
| AK224929.1 | ||||
| AK224931.1 | ||||
| AK224932.1 | ||||
| BC117102.1 | ||||
| BC117104.1 |
More Information
Set Membership: |
|

Genomic Map