Assay Details
Target Gene Details
Entrez Gene ID: | 215257 |
Gene Name: | interleukin 1 family, member 9 |
Gene Aliases: |
Il36g |
Location: |
Chr.2:24186476-24193567 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il1f9 | NM_153511.3 | NP_705731.2 | ||
| AK081783.1 | BAE43375.1 | |||
| AY071843.1 | AAL67153.1 | |||
| BC116227.1 | ||||
| BC116228.1 |
More Information
Set Membership: |
|

Genomic Map