Assay Details
Target Gene Details
Entrez Gene ID: | 16181 |
Gene Name: | interleukin 1 receptor antagonist |
Gene Aliases: |
F630041P17Rik, IL-1ra |
Location: |
Chr.2:24336860-24351491 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il1rn | NM_001039701.3 | NP_001034790.1 | ||
| NM_001159562.1 | NP_001153034.1 | |||
| NM_031167.5 | NP_112444.1 | |||
| AF001795.1 | AAC15251.1 | |||
| AK076296.1 | BAC36291.1 | |||
| AK089222.1 | BAC40797.1 | |||
| AK154601.1 | BAE32704.1 | |||
| AK155263.1 | BAE33152.1 | |||
| AK155641.1 | BAE33361.1 | |||
| AK155756.1 | BAE33421.1 | |||
| AK155798.1 | BAE33438.1 | |||
| AK155846.1 | BAE33460.1 | |||
| AK155888.1 | BAE33484.1 | |||
| AK155908.1 | BAE33497.1 | |||
| AK156034.1 | ||||
| AK157157.1 | BAE33984.1 | |||
| AK170378.1 | BAE41757.1 | |||
| AK171037.1 | BAE42202.1 | |||
| AK171385.1 | BAE42426.1 | |||
| BC042532.1 | AAH42532.1 | |||
| BY206809.1 | ||||
| M57525.1 | AAA39278.1 | |||
| M64404.1 | 1 | 62 | AAA39277.1 | |
| M74294.1 | AAA39309.1 |
More Information
Set Membership: |
|

Genomic Map