Assay Details
Target Gene Details
Entrez Gene ID: | 16181 |
Gene Name: | interleukin 1 receptor antagonist |
Gene Aliases: |
F630041P17Rik, IL-1ra |
Location: |
Chr.2:24336860-24351491 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il1rn | NM_031167.5 | NP_112444.1 | ||
| AF001795.1 | AAC15251.1 | |||
| AK076296.1 | BAC36291.1 | |||
| AK170378.1 | BAE41757.1 | |||
| BC042532.1 | AAH42532.1 |
More Information
Set Membership: |
|

Genomic Map