Assay Details
Target Gene Details
Entrez Gene ID: | 228550 |
Gene Name: | inositol 1,4,5-trisphosphate 3-kinase A |
Gene Aliases: |
- |
Location: |
Chr.2:119742239-119751253 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itpka | NM_146125.2 | NP_666237.1 | ||
| BC027291.1 | AAH27291.1 |
More Information
Set Membership: |
|

Genomic Map