Assay Details
Target Gene Details
Entrez Gene ID: | 241447 |
Gene Name: | ceramide synthase 6 |
Gene Aliases: |
4732462C07Rik, AW544719, Lass6, T1L |
Location: |
Chr.2:68861137-69114282 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cers6 | NM_172856.3 | NP_766444.1 | ||
| AK028849.1 | BAC26153.1 | |||
| AK135798.1 | ||||
| AK151848.1 | BAE30739.1 | |||
| AK152417.1 | BAE31202.1 | |||
| BC057629.1 | AAH57629.1 |

Genomic Map