Assay Details
Target Gene Details
Entrez Gene ID: | 241447 |
Gene Name: | ceramide synthase 6 |
Gene Aliases: |
4732462C07Rik, AW544719, Lass6, T1L |
Location: |
Chr.2:68861137-69114282 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cers6 | NM_172856.3 | 9 | 1000 | NP_766444.1 |
| AK028849.1 | 9 | 1001 | BAC26153.1 | |
| AK135798.1 | 9 | 1102 | ||
| AK151848.1 | 9 | 1089 | BAE30739.1 | |
| AK152417.1 | 9 | 1070 | BAE31202.1 | |
| BC057629.1 | 9 | 1043 | AAH57629.1 |

Genomic Map