Assay Details
Target Gene Details
Entrez Gene ID: | 16538 |
Gene Name: | K+ voltage-gated channel, subfamily S, 1 |
Gene Aliases: |
Kv9.1 |
Location: |
Chr.2:164163619-164171113 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcns1 | NM_008435.2 | NP_032461.2 | ||
| AF008573.1 | AAB72050.1 | |||
| BY127291.1 |
More Information
Set Membership: |
|

Genomic Map