Assay Details
Target Gene Details
Entrez Gene ID: | 100043609 |
Gene Name: | predicted gene 14207 |
Gene Aliases: |
Dll4, OTTMUSG00000015946 |
Location: |
Chr.2:119321199-119326197 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm14207 | NR_030683.1 | 2 | 337 | |
| AK039958.1 | 2 | 338 | BAE20559.1 |

Genomic Map