Assay Details
Target Gene Details
Entrez Gene ID: | 99010 |
Gene Name: | lysophosphatidylcholine acyltransferase 4 |
Gene Aliases: |
AI505034, Agpat7, Aytl3, LPEAT2 |
Location: |
Chr.2:112239841-112247111 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lpcat4 | NM_207206.2 | NP_997089.1 | ||
| BC068131.1 | AAH68131.1 | |||
| BC080829.1 | AAH80829.1 |
More Information
Set Membership: |
|

Genomic Map