Assay Details
Target Gene Details
Entrez Gene ID: | 17183 |
Gene Name: | matrilin 4 |
Gene Aliases: |
matrilin-4 |
Location: |
Chr.2:164389393-164405160 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Matn4 | NM_001252563.1 | NP_001239492.1 | ||
| NM_013592.4 | NP_038620.2 | |||
| AJ006140.1 | CAA06889.1 | |||
| AK041073.1 | ||||
| BC036558.1 | AAH36558.1 |
More Information
Set Membership: |
|

Genomic Map