Assay Details
Target Gene Details
Entrez Gene ID: | 228607 |
Gene Name: | mitochondrial antiviral signaling protein |
Gene Aliases: |
D430028G21Rik, IPS-1, Visa, cardif |
Location: |
Chr.2:131234063-131248025 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mavs | NM_001206382.1 | 6 | 757 | NP_001193311.1 |
| NM_001206383.1 | 4 | 491 | NP_001193312.1 | |
| NM_001206385.1 | 5 | 666 | NP_001193314.1 | |
| NM_144888.2 | 5 | 894 | NP_659137.1 | |
| AK028421.1 | 5 | 665 | BAC25940.1 | |
| AK085038.1 | 5 | 853 | ||
| AK153748.1 | 6 | 759 | BAE32168.1 | |
| AX775773.1 | 5 | 643 | CAE11586.1 | |
| BC020006.1 | 5 | 626 | AAH20006.1 | |
| BC025825.1 | 5 | 819 | AAH25825.1 | |
| BC031352.1 | 5 | 803 | AAH31352.1 | |
| BC037391.1 | 4 | 605 | AAH37391.1 | |
| DQ167127.1 | 4 | 543 | ABA54891.1 | |
| DQ174271.1 | 4 | 543 | AAZ80418.1 |

Genomic Map