Assay Details
Target Gene Details
Entrez Gene ID: | 17076 |
Gene Name: | lymphocyte antigen 75 |
Gene Aliases: |
CD205, DEC-205, DEC205 |
Location: |
Chr.2:60293760-60383231 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 27 - Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ly75 | NM_013825.3 | NP_038853.2 | ||
| AF395445.1 | AAK81722.1 |

Genomic Map