Assay Details
Target Gene Details
Entrez Gene ID: | 227738 |
Gene Name: | leucine rich repeat and sterile alpha motif containing 1 |
Gene Aliases: |
- |
Location: |
Chr.2:32925215-32961654 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lrsam1 | NM_199302.2 | NP_955006.1 | ||
| BC049146.1 | AAH49146.1 |

Genomic Map