Assay Details
Target Gene Details
Entrez Gene ID: | 227619 |
Gene Name: | mannosidase, alpha, class 1B, member 1 |
Gene Aliases: |
E430019H13Rik, ERMan1, Gm108, MANA-ER |
Location: |
Chr.2:25332743-25352213 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Man1b1 | NM_001029983.2 | 4 | 577 | NP_001025154.1 |
| AK043541.1 | 4 | 558 | BAC31574.1 | |
| BC006645.1 | 2 | 193 | AAH06645.1 | |
| BC138550.1 | 4 | 533 | ||
| BC138551.1 | 4 | 533 | ||
| BY732333.1 | 4 | 573 |
More Information
Set Membership: |
|

Genomic Map