Assay Details
Target Gene Details
Entrez Gene ID: | 99045 |
Gene Name: | mitochondrial ribosomal protein S26 |
Gene Aliases: |
AI648866, GI008, MRP-S13, MRP-S26, Rpms13 |
Location: |
Chr.2:130563757-130565394 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrps26 | NM_207207.1 | NP_997090.1 | ||
| AK131626.1 | 2 | 450 | ||
| AK169837.1 | ||||
| AK213019.1 | ||||
| BC048517.1 | AAH48517.1 |
More Information
Set Membership: |
|

Genomic Map