Assay Details
Target Gene Details
Entrez Gene ID: | 118451 |
Gene Name: | mitochondrial ribosomal protein S2 |
Gene Aliases: |
1500019M10Rik |
Location: |
Chr.2:28465956-28471177 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrps2 | NM_001166031.2 | NP_001159503.2 | ||
| NM_080452.3 | NP_536700.3 | |||
| AB055389.1 | BAB62530.1 | |||
| AK051184.1 | BAC34550.1 | |||
| AK077219.1 | BAC36691.1 | |||
| AK154131.1 | BAE32396.1 | |||
| AK156412.1 | ||||
| AK160129.1 | ||||
| AK165956.1 | BAE38484.1 | |||
| AK171699.1 | ||||
| AK172120.1 | 2 | 223 | ||
| AK172432.1 | ||||
| BB869096.1 | ||||
| BC060503.1 | AAH60503.1 |
More Information
Set Membership: |
|

Genomic Map