Assay Details
Target Gene Details
Entrez Gene ID: | 66559 |
Gene Name: | methionyl aminopeptidase type 1D (mitochondrial) |
Gene Aliases: |
2310066F24Rik, 3110033D18Rik, AV117938, MAP 1D, Map1d, MetAP 1D, Metapl1 |
Location: |
Chr.2:71453319-71525196 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Metap1d | NM_025633.3 | NP_079909.1 | ||
| AK005241.1 | BAB23899.1 | |||
| AK010067.1 | BAB26680.1 |
More Information
Set Membership: |
|

Genomic Map