Assay Details
Target Gene Details
Entrez Gene ID: | 50884 |
Gene Name: | NCK-associated protein 1 |
Gene Aliases: |
C79304, H19, Hem-2, Hem2, Nap1, mKIAA0587, mh19, p125Nap1 |
Location: |
Chr.2:80500512-80581259 on Build GRCm38 |
Assay Gene Location: | Within Exon 32 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nckap1 | NM_001290745.1 | 32 | 3568 | NP_001277674.1 |
| NM_016965.3 | 31 | 3550 | NP_058661.1 | |
| AJ534525.1 | 31 | 3333 | CAD58932.1 | |
| AK122318.1 | 31 | 4896 | BAC65600.1 | |
| AK143049.1 | 8 | 708 | BAE25258.1 | |
| BC003962.1 | 11 | 1072 | AAH03962.1 | |
| BC127067.1 | 32 | 3350 |

Genomic Map