Assay Details
Target Gene Details
Entrez Gene ID: | 78049 |
Gene Name: | RIKEN cDNA 4930573O16 gene |
Gene Aliases: |
- |
Location: |
Chr.2:52291002-52315240 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4930573O16Rik | NR_040620.1 | |||
| AK019812.1 |
Target Gene Details
Entrez Gene ID: | 17996 |
Gene Name: | nebulin |
Gene Aliases: |
AI595938 |
Location: |
Chr.2:52136640-52338798 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Neb | NM_010889.1 | NP_035019.1 | ||
| AF203898.1 | AAF59978.1 |
More Information
Set Membership: |
|

Genomic Map