Assay Details
Target Gene Details
Entrez Gene ID: | 17957 |
Gene Name: | N-ethylmaleimide sensitive fusion protein attachment protein beta |
Gene Aliases: |
Brp14, E161, I47, SNARE, b-SNAP |
Location: |
Chr.2:148693985-148732420 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Napb | NM_019632.3 | NP_062606.1 | ||
| AK083184.1 | BAC38798.1 | |||
| BC038362.1 | AAH38362.1 | |||
| BC049874.1 | AAH49874.1 | |||
| X61455.1 | CAA43695.1 |
More Information
Set Membership: |
|

Genomic Map