Assay Details
Target Gene Details
Entrez Gene ID: | 17996 |
Gene Name: | nebulin |
Gene Aliases: |
AI595938 |
Location: |
Chr.2:52136640-52338798 on Build GRCm38 |
Assay Gene Location: | Within Exon 154 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Neb | NM_010889.1 | 154 | 21403 | NP_035019.1 |
| AA915697.1 | 2 | 136 | ||
| AF203899.1 | 18 | 1832 | AAF59979.1 | |
| CR758733.1 |

Genomic Map