Assay Details
Target Gene Details
Entrez Gene ID: | 329416 |
Gene Name: | nitric oxide synthase trafficker |
Gene Aliases: |
Daip2, mDaIP2 |
Location: |
Chr.2:69135800-69189330 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nostrin | NM_181547.3 | 14 | 1330 | NP_853525.3 |
| AJ564264.1 | 14 | 1240 | CAD91927.1 | |
| AY280963.1 | 14 | 1283 | AAP32311.1 | |
| BC069942.1 | 14 | 1307 | AAH69942.1 |

Genomic Map