Assay Details
Target Gene Details
Entrez Gene ID: | 18128 |
Gene Name: | notch 1 |
Gene Aliases: |
9930111A19Rik, Mis6, N1, Tan1, lin-12 |
Location: |
Chr.2:26457902-26503822 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 27 - Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Notch1 | NM_008714.3 | NP_032740.3 | ||
| AB101625.1 | ||||
| AB101626.1 | ||||
| AB101627.1 | ||||
| AB101628.1 | ||||
| AB101634.1 | ||||
| AF508809.1 | AAM28905.1 | |||
| AK054089.1 | ||||
| AK090118.1 | ||||
| AK154528.1 | BAE32653.1 | |||
| AK157475.1 | BAE34095.1 | |||
| BC138441.1 | ||||
| BC138442.1 | ||||
| L02613.2 | AAK14898.1 | |||
| Z11886.1 | CAA77941.1 |

Genomic Map