Assay Details
Target Gene Details
Entrez Gene ID: | 18146 |
Gene Name: | neural proliferation, differentiation and control 1 |
Gene Aliases: |
AI314472, NPDC-1 |
Location: |
Chr.2:25403050-25409494 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Npdc1 | NM_008721.4 | NP_032747.2 | ||
| AK146466.1 | BAE27193.1 | |||
| AK210832.1 | ||||
| BC003320.1 | AAH03320.1 | |||
| BC023887.1 | AAH23887.1 | |||
| L03814.1 | AAA39836.1 | |||
| X67209.1 | CAA47648.1 |

Genomic Map