Assay Details
Target Gene Details
Entrez Gene ID: | 18019 |
Gene Name: | nuclear factor of activated T cells, cytoplasmic, calcineurin dependent 2 |
Gene Aliases: |
AI607462, NF-ATc2, NF-ATp, NFAT1, NFAT1-D, Nfatp |
Location: |
Chr.2:168476410-168601657 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nfatc2 | NM_001037178.2 | NP_001032255.1 | ||
| NM_001291168.1 | NP_001278097.1 | |||
| NM_001291169.1 | NP_001278098.1 | |||
| NM_001291170.1 | NP_001278099.1 | |||
| NM_001291171.1 | NP_001278100.1 | |||
| NM_001291172.1 | NP_001278101.1 | |||
| NM_001291173.1 | NP_001278102.1 | |||
| NM_001291174.1 | NP_001278103.1 | |||
| NR_111897.1 | ||||
| AK044489.1 | ||||
| AK083119.1 | BAC38768.1 | |||
| EU887583.1 | ||||
| EU887585.1 | ||||
| EU887589.1 | ||||
| EU887591.1 | ||||
| EU887595.1 | ||||
| EU887597.1 | ||||
| EU887601.1 | ||||
| EU887603.1 |
More Information
Set Membership: |
|

Genomic Map