Assay Details
Target Gene Details
Entrez Gene ID: | 14536 |
Gene Name: | nuclear receptor subfamily 6, group A, member 1 |
Gene Aliases: |
1700113M01Rik, Gcnf, NCNF, RTR |
Location: |
Chr.2:38723369-38927688 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nr6a1 | NM_001159548.1 | 8 | 1225 | NP_001153020.1 |
| NM_001159549.1 | 8 | 1222 | NP_001153021.1 | |
| NM_010264.4 | 10 | 1533 | NP_034394.1 | |
| AF390896.1 | 11 | 1678 | AAM55193.1 | |
| CK638822.1 | 4 | 622 | ||
| CN532747.1 | 1 | 46 | ||
| U09563.1 | 10 | 1533 | AAA64746.1 | |
| U14666.1 | 10 | 1492 | AAA92682.1 |

Genomic Map