Assay Details
Target Gene Details
Entrez Gene ID: | 14536 |
Gene Name: | nuclear receptor subfamily 6, group A, member 1 |
Gene Aliases: |
1700113M01Rik, Gcnf, NCNF, RTR |
Location: |
Chr.2:38723369-38927688 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nr6a1 | NM_001159548.1 | NP_001153020.1 | ||
| NM_001159549.1 | NP_001153021.1 | |||
| NM_010264.4 | NP_034394.1 | |||
| AF390896.1 | AAM55193.1 | |||
| AK141017.1 | ||||
| CN537360.1 | ||||
| CN537407.1 | ||||
| CN538225.1 | ||||
| U09563.1 | AAA64746.1 | |||
| U14666.1 | AAA92682.1 |

Genomic Map