Assay Details
Target Gene Details
Entrez Gene ID: | 386649 |
Gene Name: | NSFL1 (p97) cofactor (p47) |
Gene Aliases: |
Munc-18c, Stxbp3a, p47 |
Location: |
Chr.2:151494182-151511310 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nsfl1c | NM_001291074.1 | NP_001278003.1 | ||
| NM_198326.3 | NP_938085.1 | |||
| AK013024.1 | BAB28604.1 | |||
| AK039136.1 | BAC30250.1 | |||
| AK137088.1 | BAE23234.1 | |||
| AK181317.1 | ||||
| BC050936.1 | AAH50936.1 | |||
| BC106101.1 | AAI06102.1 | |||
| CJ152490.1 |
More Information
Set Membership: |
|

Genomic Map