Assay Details
Target Gene Details
Entrez Gene ID: | 18041 |
Gene Name: | nitrogen fixation gene 1 (S. cerevisiae) |
Gene Aliases: |
AA987187, m-Nfs1, m-Nfsl |
Location: |
Chr.2:156123637-156144186 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nfs1 | NM_010911.2 | 3 | 326 | NP_035041.2 |
| AJ222660.1 | 3 | 325 | CAA10916.1 | |
| AK003786.1 | 3 | 320 | ||
| AK005724.1 | 3 | 315 | ||
| AK075575.1 | 3 | 324 | BAC35831.1 | |
| AK084831.1 | 3 | 295 | ||
| AK132838.1 | 3 | 327 | BAE21386.1 | |
| AK148527.1 | 3 | 327 | BAE28601.1 | |
| AK159828.1 | 3 | 315 | BAE35407.1 | |
| AK188011.1 | ||||
| BC010586.1 | 3 | 304 | AAH10586.1 |
More Information
Set Membership: |
|

Genomic Map