Assay Details
Target Gene Details
Entrez Gene ID: | 18041 |
Gene Name: | nitrogen fixation gene 1 (S. cerevisiae) |
Gene Aliases: |
AA987187, m-Nfs1, m-Nfsl |
Location: |
Chr.2:156123637-156144186 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nfs1 | NM_010911.2 | NP_035041.2 | ||
| AJ222660.1 | CAA10916.1 | |||
| AK005724.1 | ||||
| AK075575.1 | BAC35831.1 | |||
| AK132838.1 | BAE21386.1 | |||
| AK148527.1 | BAE28601.1 | |||
| AK159828.1 | BAE35407.1 | |||
| AK183689.1 | ||||
| AK206231.1 | ||||
| AK206381.1 | ||||
| AK206572.1 | ||||
| AK207148.1 | ||||
| AK207847.1 | ||||
| AK207895.1 | ||||
| AK211411.1 | ||||
| AK211628.1 | ||||
| AK212905.1 | ||||
| AK216163.1 | ||||
| AK220082.1 | ||||
| BC010586.1 | AAH10586.1 |
More Information
Set Membership: |
|

Genomic Map