Assay Details
Target Gene Details
Entrez Gene ID: | 58237 |
Gene Name: | Na+/K+ transporting ATPase interacting 4 |
Gene Aliases: |
AB030182, C030019F02Rik |
Location: |
Chr.2:180934769-180954888 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nkain4 | NM_001141933.1 | 1 | 70 | NP_001135405.1 |
| NM_021426.4 | 1 | 70 | NP_067401.3 | |
| AB030182.1 | BAA92746.1 | |||
| AK005262.1 | BAB23914.1 | |||
| AK007632.1 | BAB25149.1 | |||
| AK021096.1 | 1 | 65 | BAB32298.1 | |
| BC061107.1 | AAH61107.1 | |||
| BC089330.1 | AAH89330.1 | |||
| CK625327.1 | 1 | 70 | ||
| EF058055.1 | ||||
| EF058056.1 | ||||
| EF058057.1 |

Genomic Map