Assay Details
Target Gene Details
Entrez Gene ID: | 18534 |
Gene Name: | phosphoenolpyruvate carboxykinase 1, cytosolic |
Gene Aliases: |
AI265463, PEPCK, Pck-1 |
Location: |
Chr.2:173153073-173159254 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pck1 | NM_011044.2 | 3 | 452 | NP_035174.1 |
| AK028046.1 | 3 | 453 | BAC25718.1 | |
| AK030327.1 | 3 | 452 | BAC26903.1 | |
| AK080261.1 | 2 | 775 | ||
| AK133496.1 | 3 | 453 | BAE21687.1 | |
| AK149525.1 | 3 | 453 | BAE28938.1 | |
| BC037629.1 | 3 | 447 | AAH37629.1 |
More Information
Set Membership: |
|

Genomic Map